4-(Difluormethoxy)zimtsäure

Lieferant: SIA ENAMINE

EN30005280.1G
ENAMEN30005280.1GEA 0 EUR
ENAMEN30005280.1G
4-(Difluormethoxy)zimtsäure
4-(Difluormethoxy)zimtsäure
Formel: C₁₀H₈F₂O₃
MDL: MFCD01222822
CAS-Nummer: 103962-07-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR