4-(Benzyloxy)-3-brombenzaldehyd

Lieferant: SIA ENAMINE

EN30005378.1G
ENAMEN30005378.1GEA 0 EUR
ENAMEN30005378.1G
4-(Benzyloxy)-3-brombenzaldehyd
4-(Benzyloxy)-3-brombenzaldehyd
Formel: C₁₄H₁₁BrO₂
MDL: MFCD02629356
CAS-Nummer: 69455-12-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR