1-(2-(Trifluormethyl)phenyl)thioharnstoff

Lieferant: SIA ENAMINE

Synonyme: N-[2-(Trifluoromethyl)phenyl]thiourea

EN30005675.1G
ENAMEN30005675.1GEA 0 EUR
ENAMEN30005675.1G
1-(2-(Trifluormethyl)phenyl)thioharnstoff
1-(2-(Trifluormethyl)phenyl)thioharnstoff
Formel: C₈H₇F₃N₂S
Molecular Weight: 220,22 g/mol
MDL: MFCD00041161
CAS-Nummer: 1736-71-6
EINECS: 217-084-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR