Ethyl-2-amino-5-methyl-4-phenyl-3-thiophencarboxylat

Lieferant: SIA ENAMINE

EN30005677.1G
ENAMEN30005677.1GEA 0 EUR
ENAMEN30005677.1G
Ethyl-2-amino-5-methyl-4-phenyl-3-thiophencarboxylat
Ethyl-2-amino-5-methyl-4-phenyl-3-thiophencarboxylat
Formel: C₁₄H₁₅NO₂S
Molecular Weight: 261.34 g/mol
MDL: MFCD01763475
CAS-Nummer: 4815-37-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR