3,4-(Ethylendioxy)thiophenol

Lieferant: SIA ENAMINE

Synonyme: 2,3-Dihydrobenzo[b][1,4]dioxine-6-thiol

EN30005854.1G
ENAMEN30005854.1GEA 0 EUR
ENAMEN30005854.1G
3,4-(Ethylendioxy)thiophenol
3,4-(Ethylendioxy)thiophenol
Formel: C₈H₈O₂S
Molecular Weight: 168,212 g/mol
MDL: MFCD06201753
CAS-Nummer: 247228-28-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR