7-Isochinolinol

Lieferant: SIA ENAMINE
Achtung

Synonyme: 7-Hydroxyisoquinoline, Isoquinolin-7-ol

EN30005942.1G
ENAMEN30005942.1GEA 0 EUR
ENAMEN30005942.1G
7-Isochinolinol
7-Isochinolinol
Formel: C₉H₇NO
Molecular Weight: 145,16 g/mol
MDL: MFCD00456131
CAS-Nummer: 7651-83-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR