1-(4-Hydroxyphenyl)-1H-tetrazol

Lieferant: SIA ENAMINE

Synonyme: 4-(1H-Tetrazol-1-yl)phenol

EN30006032.1G
ENAMEN30006032.1GEA 0 EUR
ENAMEN30006032.1G
1-(4-Hydroxyphenyl)-1H-tetrazol
1-(4-Hydroxyphenyl)-1H-tetrazole
Formel: C₇H₆N₄O
Molecular Weight: 162,1500 g/mol
MDL: MFCD03490124
CAS-Nummer: 64001-11-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR