1-(4-Chlorphenyl)-2,5-dimethylpyrrol

Lieferant: SIA ENAMINE

Synonyme: 1-(4-Chlorophenyl)-2,5-dimethyl-1H-pyrrole

EN30006146.1G
ENAMEN30006146.1GEA 0 EUR
ENAMEN30006146.1G
1-(4-Chlorphenyl)-2,5-dimethylpyrrol
1-(4-Chlorophenyl)-2,5-dimethylpyrrole
Formel: C₁₂H₁₂ClN
MDL: MFCD00027420
CAS-Nummer: 5044-23-5
EINECS: 225-739-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR