3,4-Xylylessigsäure

Lieferant: SIA ENAMINE

Synonyme: 3,4-Dimethylphenylacetic acid

EN30006309.1G
ENAMEN30006309.1GEA 0 EUR
ENAMEN30006309.1G
3,4-Xylylessigsäure
3,4-Xylylessigsäure
Formel: C₁₀H₁₂O₂
Molecular Weight: 164,2 g/mol
MDL: MFCD02664684
CAS-Nummer: 17283-16-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR