1-(2,5-Dimethylphenyl)-2-thioharnstoff

Lieferant: SIA ENAMINE

EN30006326.1G
ENAMEN30006326.1GEA 0 EUR
ENAMEN30006326.1G
1-(2,5-Dimethylphenyl)-2-thioharnstoff
1-(2,5-Dimethylphenyl)-2-thioharnstoff
Formel: (C₆H₃(CH₃)₂)NC(S)NH₂
Molecular Weight: 180,27 g/mol
MDL: MFCD00041372
CAS-Nummer: 16738-19-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR