4-Methoxydiphenylamin
Lieferant: SIA ENAMINE
EN30006439.1G
ENAMEN30006439.1GEA
0
EUR
ENAMEN30006439.1G
4-Methoxydiphenylamin
4-Methoxydiphenylamin
Formel:
C₁₃H₁₃NO |
MDL:
MFCD00228649 CAS-Nummer: 1208-86-2 EINECS: 214-902-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...