3-(3-Bromphenyl)propionsäure

Lieferant: SIA ENAMINE

EN30006752.1G
ENAMEN30006752.1GEA 0 EUR
ENAMEN30006752.1G
3-(3-Bromphenyl)propionsäure
3-(3-Bromphenyl)propionsäure
Formel: C₉H₉BrO₂
MDL: MFCD01310792
CAS-Nummer: 42287-90-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR