5-Isoxazolcarbonsäure

Lieferant: SIA ENAMINE

Synonyme: Isoxazole-5-carboxylic acid

EN30006774.1G
ENAMEN30006774.1GEA 0 EUR
ENAMEN30006774.1G
5-Isoxazolcarbonsäure
5-Isoxazolecarboxylic acid
Formel: C₄H₃NO₃
Molecular Weight: 113,07 g/mol
MDL: MFCD00156151
CAS-Nummer: 21169-71-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR