2-Chlor-4-thiocyanatoanilin

Lieferant: SIA ENAMINE

EN30006825.1G
ENAMEN30006825.1GEA 0 EUR
ENAMEN30006825.1G
2-Chlor-4-thiocyanatoanilin
2-Chlor-4-thiocyanatoanilin
Formel: C₇H₅ClN₂S
MDL: MFCD04614735
CAS-Nummer: 3226-47-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR