2-(Cyanmethyl)benzoesäure

Lieferant: SIA ENAMINE

Synonyme: 2-Carboxyphenylacetonitrile

EN30006835.1G
ENAMEN30006835.1GEA 0 EUR
ENAMEN30006835.1G
2-(Cyanmethyl)benzoesäure
2-(Cyanmethyl)benzoesäure
Formel: C₉H₇NO₂
Molecular Weight: 161,16 g/mol
CAS-Nummer: 6627-91-4
EINECS: 229-602-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR