2-Fluor-4-thiocyanatoanilin

Lieferant: SIA ENAMINE

EN30006899.1G
ENAMEN30006899.1GEA 0 EUR
ENAMEN30006899.1G
2-Fluor-4-thiocyanatoanilin
2-Fluor-4-thiocyanatoanilin
Formel: C₇H₅FN₂S
MDL: MFCD04617227
CAS-Nummer: 14512-85-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR