6,7-Dimethoxy-4-chinazolon

Lieferant: SIA ENAMINE

Synonyme: 6,7-Dimethoxy-4(3H)-quinazolinone, 6,7-Dimethoxy-1H-quinazolin-4-one

EN30006937.1G
ENAMEN30006937.1GEA 0 EUR
ENAMEN30006937.1G
6,7-Dimethoxy-4-chinazolon
6,7-Dimethoxy-4-chinazolon
Formel: C₁₀H₁₀N₂O₃
Molecular Weight: 206,2 g/mol
MDL: MFCD01570147
CAS-Nummer: 13794-72-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR