4-(4-Bromphenoxy)anilin

Lieferant: SIA ENAMINE

EN30006982.1G
ENAMEN30006982.1GEA 0 EUR
ENAMEN30006982.1G
4-(4-Bromphenoxy)anilin
4-(4-Bromphenoxy)anilin
Formel: C₁₂H₁₀BrNO
CAS-Nummer: 31465-35-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR