4'-Methoxy-2-chloracetophenon

Lieferant: SIA ENAMINE

Synonyme: 2-Chloro-1-(4-methoxyphenyl)ethanone, 4-Methoxyphenacyl chloride

EN30007125.1G
ENAMEN30007125.1GEA 0 EUR
ENAMEN30007125.1G
4'-Methoxy-2-chloracetophenon
4'-Methoxy-2-chloracetophenon
Formel: C₉H₉ClO₂
MDL: MFCD00216508
CAS-Nummer: 2196-99-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR