7-Fluor-2-methylchinolin-3-carbonsäure

Lieferant: SIA ENAMINE

EN30007134.1G
ENAMEN30007134.1GEA 0 EUR
ENAMEN30007134.1G
7-Fluor-2-methylchinolin-3-carbonsäure
7-Fluor-2-methylchinolin-3-carbonsäure
Formel: C₁₁H₈FNO₂
MDL: MFCD06254735
CAS-Nummer: 879361-44-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR