5-Brom-2,3-diaminopyridin

Lieferant: SIA ENAMINE

Synonyme: 5-Bromopyridine-2,3-diyldiamine, 2,3-Diamino-5-bromopyridine

EN30007258.1G
ENAMEN30007258.1GEA 0 EUR
ENAMEN30007258.1G
5-Brom-2,3-diaminopyridin
5-Brom-2,3-diaminopyridin
Formel: C₅H₆BrN₃
Molecular Weight: 188,03 g/mol
Siedepunkt: 321 °C (1013 hPa)
MDL: MFCD00460094
CAS-Nummer: 38875-53-5
EINECS: 254-172-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR