1-(4-Aminophenyl)-1H-tetrazol

Lieferant: SIA ENAMINE

EN30007323.1G
ENAMEN30007323.1GEA 0 EUR
ENAMEN30007323.1G
1-(4-Aminophenyl)-1H-tetrazol
1-(4-Aminophenyl)-1H-tetrazole
Formel: C₇H₇N₅
MDL: MFCD02746184
CAS-Nummer: 14213-13-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR