2-Fluor-N-methylbenzylamin

Lieferant: SIA ENAMINE

Synonyme: 1-(2-Fluorophenyl)-N-methylmethanamine, (2-Fluorobenzyl)methylamine

EN30007361.1G
ENAMEN30007361.1GEA 0 EUR
ENAMEN30007361.1G
2-Fluor-N-methylbenzylamin
2-Fluor-N-methylbenzylamin
Formel: C₈H₁₀FN
Molecular Weight: 139,17 g/mol
MDL: MFCD04629633
CAS-Nummer: 399-30-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR