2,3-Dichlorzimtsäure

Lieferant: SIA ENAMINE

Synonyme: 2,3-Dichlorcinnamsäure, 3-(2,3-Dichlorphenyl)propensäure

EN30007610.1G
ENAMEN30007610.1GEA 0 EUR
ENAMEN30007610.1G
2,3-Dichlorzimtsäure
2,3-Dichlorzimtsäure
Formel: C₉H₆Cl₂O₂
Molecular Weight: 217,049 g/mol
MDL: MFCD01076352
CAS-Nummer: 20595-44-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR