3,5-Dimethylphenylhydraziniumchlorid

Lieferant: SIA ENAMINE

Synonyme: 3,5-Dimethylphenylhydrazinhydrochlorid, 1-(3,5-Dimethylphenyl)hydrazin Hydrochlorid, 3,5-Dimethylphenylhydrazin Hydrochlorid

EN30007656.1G
ENAMEN30007656.1GEA 0 EUR
ENAMEN30007656.1G
3,5-Dimethylphenylhydraziniumchlorid
3,5-Dimethylphenylhydraziniumchlorid
Formel: C₈H₁₃ClN₂
Molecular Weight: 172,66 g/mol
MDL: MFCD00052269
CAS-Nummer: 60481-36-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR