2,5-Dimethylphenylhydraziniumchlorid

Lieferant: SIA ENAMINE

Synonyme: 1-(2,5-Dimethylphenyl)hydrazin Hydrochlorid, 2,5-Dimethylphenylhydrazinhydrochlorid, 2,5-Dimethylphenylhydrazin Hydrochlorid

EN30007716.1G
ENAMEN30007716.1GEA 0 EUR
ENAMEN30007716.1G
2,5-Dimethylphenylhydraziniumchlorid
2,5-Dimethylphenylhydraziniumchlorid
Formel: C₈H₁₃ClN₂
MDL: MFCD00013382
CAS-Nummer: 56737-78-1
UN: 2811
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR