3-Hydroxy-5-methylisoxazol

Lieferant: SIA ENAMINE

Synonyme: Hymexazol

EN30007726.1G
ENAMEN30007726.1GEA 0 EUR
ENAMEN30007726.1G
3-Hydroxy-5-methylisoxazol
3-Hydroxy-5-methylisoxazole
Formel: C₄H₅NO₂
Molecular Weight: 99,09 g/mol
MDL: MFCD00144468
CAS-Nummer: 10004-44-1
EINECS: 233-000-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR