3'-Fluor-4'-morpholinoacetophenon

Lieferant: SIA ENAMINE

EN30007759.1G
ENAMEN30007759.1GEA 0 EUR
ENAMEN30007759.1G
3'-Fluor-4'-morpholinoacetophenon
3'-Fluor-4'-morpholinoacetophenon
Formel: C₁₂H₁₄FNO₂
MDL: MFCD00543914
CAS-Nummer: 189763-65-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR