2-Brom-4-thiocyanatoanilin

Lieferant: SIA ENAMINE

EN30007851.1G
ENAMEN30007851.1GEA 0 EUR
ENAMEN30007851.1G
2-Brom-4-thiocyanatoanilin
2-Brom-4-thiocyanatoanilin
Formel: C₇H₅BrN₂S
MDL: MFCD04618254
CAS-Nummer: 7493-98-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR