4-Hydroxy-3-methoxybenzonitril

Lieferant: SIA ENAMINE

EN30007946.1G
ENAMEN30007946.1GEA 0 EUR
ENAMEN30007946.1G
4-Hydroxy-3-methoxybenzonitril
4-Hydroxy-3-methoxybenzonitril
Formel: C₈H₇NO₂
MDL: MFCD00001820
CAS-Nummer: 4421-08-3
EINECS: 224-589-0
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR