4-Fluor-N-methylbenzylamin

Lieferant: SIA ENAMINE

Synonyme: N-Methyl-4-fluorobenzylamine

EN30007965.1G
ENAMEN30007965.1GEA 0 EUR
ENAMEN30007965.1G
4-Fluor-N-methylbenzylamin
4-Fluor-N-methylbenzylamin
Formel: C₈H₁₀FN
Molecular Weight: 139.17 g/mol
MDL: MFCD03839848
CAS-Nummer: 405-66-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR