3-Amino-5-phenyl-1H-pyrazol

Lieferant: SIA ENAMINE

Synonyme: 3-Amino-5-phenylpyrazole

EN30007991.1G
ENAMEN30007991.1GEA 0 EUR
ENAMEN30007991.1G
3-Amino-5-phenyl-1H-pyrazol
3-Amino-5-phenyl-1H-pyrazole
Formel: C₉H₉N₃
Molecular Weight: 159,19 g/mol
MDL: MFCD00191749
CAS-Nummer: 1572-10-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR