2-Hydrazinobenzoesäure Hydrochlorid

Lieferant: SIA ENAMINE

EN30008025.1G
ENAMEN30008025.1GEA 0 EUR
ENAMEN30008025.1G
2-Hydrazinobenzoesäure Hydrochlorid
2-Hydrazinobenzoesäure Hydrochlorid
Formel: C₇H₉ClN₂O₂
MDL: MFCD00012931
CAS-Nummer: 52356-01-1
EINECS: 257-869-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR