4-Chlorphenylhydraziniumchlorid

Lieferant: SIA ENAMINE

EN30008030.1G
ENAMEN30008030.1GEA 0 EUR
ENAMEN30008030.1G
4-Chlorphenylhydraziniumchlorid
4-Chlorphenylhydraziniumchlorid
Formel: C₆H₈Cl₂N₂
MDL: MFCD00012943
CAS-Nummer: 1073-70-7
EINECS: 214-030-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR