2-Brom-4-nitroanilin
Lieferant: SIA ENAMINE
EN30008034.1G
ENAMEN30008034.1GEA
0
EUR
ENAMEN30008034.1G
2-Brom-4-nitroanilin
2-Brom-4-nitroanilin
Formel:
C₆H₅BrN₂O₂ |
MDL:
MFCD00025152 CAS-Nummer: 13296-94-1 EINECS: 236-318-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...