4-(Trifluormethyl)anthranilsäure

Lieferant: SIA ENAMINE

EN30008036.1G
ENAMEN30008036.1GEA 0 EUR
ENAMEN30008036.1G
4-(Trifluormethyl)anthranilsäure
4-(Trifluormethyl)anthranilsäure
Formel: C₈H₆F₃NO₂
Molecular Weight: 205,14 g/mol
MDL: MFCD00519334
CAS-Nummer: 402-13-1
EINECS: 206-937-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR