2,4-Dimethylphenylhydraziniumchlorid

Lieferant: SIA ENAMINE

Synonyme: 2,4-Dimethylphenylhydrazin Hydrochlorid, 1-(2,4-Dimethylphenyl)hydrazin Hydrochlorid, 2,4-Dimethylphenylhydrazinhydrochlorid

EN30008047.1G
ENAMEN30008047.1GEA 0 EUR
ENAMEN30008047.1G
2,4-Dimethylphenylhydraziniumchlorid
2,4-Dimethylphenylhydraziniumchlorid
Formel: C₈H₁₃ClN₂
Molecular Weight: 172,66 g/mol
MDL: MFCD00013381
CAS-Nummer: 60480-83-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR