2-Chlorphenylhydraziniumchlorid

Lieferant: SIA ENAMINE

Synonyme: 2-Chlorphenylhydrazin Hydrochlorid, 2-Chlorphenylhydrazinhydrochlorid, 1-(2-Chlorphenyl)hydrazin Hydrochlorid

EN30008048.1G
ENAMEN30008048.1GEA 0 EUR
ENAMEN30008048.1G
2-Chlorphenylhydraziniumchlorid
2-Chlorphenylhydraziniumchlorid
Formel: C₆H₈Cl₂N₂
Molecular Weight: 179,05 g/mol
MDL: MFCD00012928
CAS-Nummer: 41052-75-9
EINECS: 255-194-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR