3,6-Dichlorpicolinsäure

Lieferant: SIA ENAMINE

Synonyme: 3,6-Dichloropyridine-2-carboxylic acid

EN30008124.1G
ENAMEN30008124.1GEA 0 EUR
ENAMEN30008124.1G
3,6-Dichlorpicolinsäure
3,6-Dichlorpicolinsäure
Formel: C₆H₃Cl₂NO₂
Molecular Weight: 192,00 g/mol
MDL: MFCD00078655
CAS-Nummer: 1702-17-6
EINECS: 216-935-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR