2-Amino-3-thiophencarbonitril

Lieferant: SIA ENAMINE

EN30008146.1G
ENAMEN30008146.1GEA 0 EUR
ENAMEN30008146.1G
2-Amino-3-thiophencarbonitril
2-Amino-3-thiophencarbonitril
Formel: C₅H₄N₂S
MDL: MFCD00706298
CAS-Nummer: 4651-82-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR