3-(Benzyloxy)benzoesäure

Lieferant: SIA ENAMINE

Synonyme: 3-Benzyloxybenzoic acid

EN30008215.1G
ENAMEN30008215.1GEA 0 EUR
ENAMEN30008215.1G
3-(Benzyloxy)benzoesäure
3-(Benzyloxy)benzoesäure
Formel: C₁₄H₁₂O₃
Molecular Weight: 228,25 g/mol
MDL: MFCD00527901
CAS-Nummer: 69026-14-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR