6-Methoxy-2-methylbenzothiazol

Lieferant: SIA ENAMINE

EN30008349.1G
ENAMEN30008349.1GEA 0 EUR
ENAMEN30008349.1G
6-Methoxy-2-methylbenzothiazol
6-Methoxy-2-methylbenzothiazole
Formel: C₉H₉NOS
MDL: MFCD00005795
CAS-Nummer: 2941-72-2
EINECS: 220-931-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR