Ethyl α-chlorphenylacetat

Lieferant: SIA ENAMINE

Synonyme: Ethyl 2-chloro-2-phenylacetate

EN30008394.1G
ENAMEN30008394.1GEA 0 EUR
ENAMEN30008394.1G
Ethyl α-chlorphenylacetat
Ethyl α-chlorphenylacetat
Formel: C₁₀H₁₁ClO₂
Molecular Weight: 198,6461 g/mol
MDL: MFCD00013675
CAS-Nummer: 4773-33-5
EINECS: 225-315-2
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR