3',4'-Dimethylbenzophenon-2-carbonsäure
Lieferant: SIA ENAMINE
EN30008550.1G
ENAMEN30008550.1GEA
0
EUR
ENAMEN30008550.1G
3',4'-Dimethylbenzophenon-2-carbonsäure
3',4'-Dimethylbenzophenon-2-carbonsäure
Formel:
C₁₆H₁₄O₃ |
CAS-Nummer:
2159-42-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...