5-Chlor-2-nitropyridin

Lieferant: SIA ENAMINE

EN30008929.1G
ENAMEN30008929.1GEA 0 EUR
ENAMEN30008929.1G
5-Chlor-2-nitropyridin
5-Chlor-2-nitropyridin
Formel: C₅H₃ClN₂O₂
Molecular Weight: 158,54 g/mol
MDL: MFCD03092916
CAS-Nummer: 52092-47-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR