N-(4-Fluorophenyl)rhodanin

Lieferant: SIA ENAMINE

EN30008963.1G
ENAMEN30008963.1GEA 0 EUR
ENAMEN30008963.1G
N-(4-Fluorophenyl)rhodanin
N-(4-Fluorophenyl)rhodanin
Formel: C₉H₆FNOS₂
CAS-Nummer: 387-27-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR