2-Brombenzolsulfonylchlorid
Lieferant: SIA ENAMINE
Synonyme:
2-Brombenzolsulfochlorid, 2-Brombenzolsulfonsäurechlorid, 2-Brombenzol-1-sulfonylchlorid
EN30009049.1G
ENAMEN30009049.1GEA
0
EUR
ENAMEN30009049.1G
2-Brombenzolsulfonylchlorid
2-Brombenzolsulfonylchlorid
Formel:
C₆H₄BrClO₂S Molecular Weight: 255,52 g/mol Siedepunkt: 127…128 °C (3,6 mmHg) Schmelzpunkt: 51 °C |
MDL:
MFCD00051971 CAS-Nummer: 2905-25-1 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...