5-Brom-8-nitroisochinolin

Lieferant: SIA ENAMINE

EN30009137.1G
ENAMEN30009137.1GEA 0 EUR
ENAMEN30009137.1G
5-Brom-8-nitroisochinolin
5-Bromo-8-nitroisoquinoline
Formel: C₉H₅BrN₂O₂
MDL: MFCD02091227
CAS-Nummer: 63927-23-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR