4-Brom-2,6-dichlorphenol

Lieferant: SIA ENAMINE

EN30009239.1G
ENAMEN30009239.1GEA 0 EUR
ENAMEN30009239.1G
4-Brom-2,6-dichlorphenol
4-Brom-2,6-dichlorphenol
Formel: C₆H₃BrCl₂O
MDL: MFCD00019996
CAS-Nummer: 3217-15-0
EINECS: 221-740-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR