3-Propoxyphenol

Lieferant: SIA ENAMINE

Synonyme: 1-Hydroxy-2-propoxybenzene

EN30009246.1G
ENAMEN30009246.1GEA 0 EUR
ENAMEN30009246.1G
3-Propoxyphenol
3-Propoxyphenol
Formel: C₉H₁₂O₂
Molecular Weight: 152,19 g/mol
MDL: MFCD06655033
CAS-Nummer: 6280-96-2
EINECS: 228-484-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR